Learn More
SOMNI Scientific™ Pole Mount Dual E-Cylinder Manifold
SDP
Catalog No. 04355317 Shop All Somni Scientific Products
Change view
Click to view available options
Animal:
All Animals
Catalog No. Animal
04-355-317 All Animals
1 options
Catalog No. 04-355-317 Supplier SOMNI Scientific™ Supplier No. RE20104
Only null left
Add to Cart
Add to Cart
The SOMNI Pole Mount Dual E-cylinder Manifold can be used with a single or two 'E' size oxygen cylinders. The incorporated check valves prevent leakage when one cylinder is disconnected. The manifold is compact, durable and easy to install on machine poles up to 2 in. diameter.
  • Compact and durable
  • Accommodates 1 or 2 e-cylinders
  • Preset regulator
  • Incorporated check valves
  • Install on pole up to 2 in. diameter
  • Hose, wrench and mounting hardware included
Specifications
Autoclavable Not Autoclavable
Animal All Animals
For Use With (Application) Animal Oxygen Supply
Type E-Cylinder Manifold
Disposable Reusable
Sterility Non-sterile