Learn More
Grainger Pole Tamper 1300 bpm L 60 in
Catalog No. 502766284
Encompass_Preferred
The Grainger 16V992 is a 3M Scotch-Brite General Purpose Hand Pad, 6" x 9", Very Fine grade. It features an open web construction and is made of high-quality fibers for cleaning, scouring, and finishing applications. This abrasive hand pad has an aggressive very fine grade that is ideal for light scrubbing and finishing of metal, plastic, glass, and other surfaces. It effectively removes rust, oxidation, and light burrs without scratching. The pad measures 6 inches by 9 inches for convenient hand use. It is durable, lint-free, and resists fraying. The 3M Scotch-Brite General Purpose Hand Pad 16V992 provides versatile cleaning and finishing for industrial and commercial use.
Catalog No. 50-276-6284 Supplier Grainger Supplier No. 16V992
The Grainger 16V992 is a 3M Scotch-Brite General Purpose Hand Pad, 6" x 9", Very Fine grade. It features an open web construction and is made of high-quality fibers for cleaning, scouring, and finishing applications. This abrasive hand pad has an aggressive very fine grade that is ideal for light scrubbing and finishing of metal, plastic, glass, and other surfaces. It effectively removes rust, oxidation, and light burrs without scratching. The pad measures 6 inches by 9 inches for convenient hand use. It is durable, lint-free, and resists fraying. The 3M Scotch-Brite General Purpose Hand Pad 16V992 provides versatile cleaning and finishing for industrial and commercial use.