Learn More
Grainger Pole Tamper 1300 bpm L 72 in
Catalog No. 502765612
Encompass_Preferred
The Greenlee 72" Pole Tamper is a gas-powered compaction tool designed for tamping and compacting soil, gravel, asphalt, and other materials. It features a Honda GX35 4-stroke engine that delivers 1300 blows per minute to the tamping shoe for efficient compaction. The pole is made from fiberglass for durability and has a vibration-dampening grip on the handle for operator comfort. This 72" long pole tamper weighs 43 lbs and is ideal for compaction jobs that require an extended reach such as pipeline trenches, foundations, and landscaping projects. It can be used for both horizontal and vertical compaction.
Catalog No. 50-276-5612 Supplier Grainger Supplier No. 16V993
The Greenlee 72" Pole Tamper is a gas-powered compaction tool designed for tamping and compacting soil, gravel, asphalt, and other materials. It features a Honda GX35 4-stroke engine that delivers 1300 blows per minute to the tamping shoe for efficient compaction. The pole is made from fiberglass for durability and has a vibration-dampening grip on the handle for operator comfort. This 72" long pole tamper weighs 43 lbs and is ideal for compaction jobs that require an extended reach such as pipeline trenches, foundations, and landscaping projects. It can be used for both horizontal and vertical compaction.