Learn More
ABclonal Technology POLE2 Rabbit pAb
SDP
Catalog No. 5000113951
Encompass_Preferred
DNA polymerase epsilon, which is involved in DNA repair and replication, is composed of a large catalytic subunit and a small accessory subunit. The protein encoded by this gene represents the small subunit (B). Defects in this gene have been linked to colorectal cancer and to combined immunodeficiency.
Catalog No. 50-001-13951 Supplier ABclonal Technology Supplier No. A12842100UL
DNA polymerase epsilon, which is involved in DNA repair and replication, is composed of a large catalytic subunit and a small accessory subunit. The protein encoded by this gene represents the small subunit (B). Defects in this gene have been linked to colorectal cancer and to combined immunodeficiency.