Learn More
Aviva Systems Biology Corp POLE3 Antibody - N-terminal region (ARP33054_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33054_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POLE3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POLE3. Alternate names: p17, YBL1, CHRAC17, CHARAC17ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54862