Learn More
Aviva Systems Biology Corp POLE4 Antibody - N-terminal region (ARP39392_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39392_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLE4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POLE4. Alternate names: p12, YHHQ1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-57016