Learn More
Aviva Systems Biology Corp Polg Antibody - N-terminal region (ARP48580_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48580_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Polg. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-285-368