Learn More
Aviva Systems Biology Corp POLG2 Antibody - N-terminal region (ARP61960_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61960_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLG2. It was validated on Western Blot. . Alternate names: HP55, MTPOLB, PEOA4, POLB, POLG-BETA, POLGBApplicationWB. Species reactivity: Human
Catalog No. 50-287-521