Learn More
Aviva Systems Biology Corp POLI Antibody - N-terminal region (ARP52299_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52299_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLI. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POLI. Alternate names: RAD30B, RAD3OBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-693