Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Polidocanol | 9002-92-0 | 98.0% | 500 MG
SDP

Catalog No. 5000409977
Encompass_Preferred

Polidocanol is a sclerosing agent used successfully to treat extremity and esophageal varices and telangiectasias. It is intended for research use only.

  • Appearance: solid below 41°C, liquid above 45°C
  • Color: white to light yellow
  • Shipping: room temperature in continental US
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility in H2O: 50 mg/mL (with ultrasonic)
  • Solubility in DMSO: 50 mg/mL (with ultrasonic)
  • Solubility in PBS (in vivo): 120 mg/mL (clear solution, with ultrasonic)

Catalog No. 50-004-09977 Supplier Medchemexpress LLC Supplier No. HYB2106500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Polidocanol is a sclerosing agent used successfully to treat extremity and esophageal varices and telangiectasias. It is intended for research use only.

  • Appearance: solid below 41°C, liquid above 45°C
  • Color: white to light yellow
  • Shipping: room temperature in continental US
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility in H2O: 50 mg/mL (with ultrasonic)
  • Solubility in DMSO: 50 mg/mL (with ultrasonic)
  • Solubility in PBS (in vivo): 120 mg/mL (clear solution, with ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.