Learn More
OriGene POLINK2 PLS RABBIT IGG HRP DAB
SDP
Catalog No. NC0450402
Encompass_Preferred
2-step HRP polymer for enhanced detection of rabbit primary antibodies in IHC. Requires two 10 minutes incubation steps. D39 is Hu/Ms serum absorbed. Biotin Free. For manual or autostainer of frozen/paraffin human/mouse tissues.
Catalog No. NC0450402 Supplier OriGene Supplier No. D39110
2-step HRP polymer for enhanced detection of rabbit primary antibodies in IHC. Requires two 10 minutes incubation steps. D39 is Hu/Ms serum absorbed. Biotin Free. For manual or autostainer of frozen/paraffin human/mouse tissues.