Learn More
OriGene POLINK2 PLUS BROAD IGG HRP DAB
SDP
Catalog No. NC0444899
Encompass_Preferred
2-step HRP polymer detection for enhanced sensitivity of mouse and rabbit primary antibodies in IHC. Requires two 10 minute incubation steps. D41 is human serum absorbed. Biotin free. For manual or autostainer of frozen/paraffin human tissues.
Catalog No. NC0444899 Supplier OriGene Supplier No. D41110
2-step HRP polymer detection for enhanced sensitivity of mouse and rabbit primary antibodies in IHC. Requires two 10 minute incubation steps. D41 is human serum absorbed. Biotin free. For manual or autostainer of frozen/paraffin human tissues.