Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Polisher ST

Catalog No. 02557920 Shop All Thermo Scientific Products
Change view
Click to view available options
Flow Rate:
1 mL/min.
1020 mL/min.
16 L/min.
170 mL/min.
2.3 L/min.
25 mL/min.
340 mL/min.
4 mL/min.
Height (Metric):
4.5 cm
4.8 cm
5.9 cm
7.6 cm
8.8 cm
10.1 cm
15.2 cm
22.2 cm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Height (Metric)
02-557-920 1 mL/min. 4.8 cm
02-558-053 2.3 L/min. 7.6 cm
02-557-966 16 L/min. 22.2 cm
02-557-945 25 mL/min. 4.5 cm
02-557-834 1020 mL/min. 15.2 cm
02-557-858 170 mL/min. 10.1 cm
02-557-796 340 mL/min. 10.1 cm
02-557-881 25 mL/min. 8.8 cm
02-557-882 4 mL/min. 5.9 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
The brand will transition from 3M to Thermo Scientific
Catalog No. 02-557-920 Supplier Thermo Scientific™ Supplier No. BC1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The brand will transition from 3M to Thermo Scientific

Single-use anion exchange media combines Q-functional nonwoven and guanidinium-functional membrane layers to reduce HCP, viruses, DNA, turbidity, and other negatively charged contaminants in flow-through polishing.

  • Fully encapsulated, single-use anion exchange (AEX) product for flow-through polishing of biopharmaceutical process streams.
  • Uses a Q-functional nonwoven media layer and a guanidinium-functional membrane.
  • Reduces host cell protein (HCP), viruses and other negatively charged contaminants.
  • Q-functional nonwoven media also supports reduction of residual turbidity and DNA.
  • Operates in a salt-tolerant polishing mode compared with conventional Q-functional AEX resins.
  • Designed as a single-use alternative to downstream AEX polishing columns in suitable processes.
  • Supports polishing of therapeutic recombinant protein process streams.

Specifications

Material (Membrane) Polyamide
Pore Size 0.8 μm
Flow Rate 1 mL/min.
Model BC1
Quantity 4/Cs.
Height (Metric) 4.8 cm
Diameter (Metric) 4.3 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.