Learn More
Electron Microscopy Sciences Billiard Polishing Cloth 10" no hole, plain back, no hole
SDP
Catalog No. 50997216
Encompass_Preferred
100% virgin wool sheared pile ideal for the preparation of metallographic specimens and materials needing a fine finish. Recommended for intermediate and final polishing steps.
Diameter: 10"
Backing: Plain
Type: no hole
Catalog No. 50-997-216 Supplier Electron Microscopy Sciences Supplier No. 5030010
100% virgin wool sheared pile ideal for the preparation of metallographic specimens and materials needing a fine finish. Recommended for intermediate and final polishing steps.
Diameter: 10"
Backing: Plain
Type: no hole