Learn More
Electron Microscopy Sciences Billiard Polishing Cloth 8" no hole, plain back, no hole
SDP
Catalog No. 50997215
Encompass_Preferred
100% virgin wool sheared pile ideal for the preparation of metallographic specimens and materials needing a fine finish. Recommended for intermediate and final polishing steps.
Diameter: 8"
Backing: PSA
Type: no hole
Catalog No. 50-997-215 Supplier Electron Microscopy Sciences Supplier No. 5030008
100% virgin wool sheared pile ideal for the preparation of metallographic specimens and materials needing a fine finish. Recommended for intermediate and final polishing steps.
Diameter: 8"
Backing: PSA
Type: no hole