Learn More
Electron Microscopy Sciences Glenco Flocked Polishing Cloth PSA, 2.5" centered hole, 8" diameter
SDP
Catalog No. 50997223
Encompass_Preferred
This flocked cloth is made by binding a rayon flock to a woven cotton backing; it is the most popular in our line. It is excellent for Final polishing when used in conjunction with diamond or alumina polishing compounds. Available with Pressure Sensitive Adhesive.
Type: 2.5" centered hole
Diameter: 8"
Catalog No. 50-997-223 Supplier Electron Microscopy Sciences Supplier No. 5031815
This flocked cloth is made by binding a rayon flock to a woven cotton backing; it is the most popular in our line. It is excellent for Final polishing when used in conjunction with diamond or alumina polishing compounds. Available with Pressure Sensitive Adhesive.
Type: 2.5" centered hole
Diameter: 8"