Learn More
Electron Microscopy Sciences Spec-Cloth Spec-Cloth Polishing Cloth, PSA, 2.5" centered hole, 8"
SDP
Catalog No. 50997225
Encompass_Preferred
A long napped rayon flock with a plastic barrier. This is a finer version of our Glenco cloth especially designed for superior final polishing. For best results, use with our diamond polishing compound. Pressue Sensitive Adhesive back and 2 1/2" hole.
Type: 2.5" centered hole
Diameter: 8"
Catalog No. 50-997-225 Supplier Electron Microscopy Sciences Supplier No. 5032015
A long napped rayon flock with a plastic barrier. This is a finer version of our Glenco cloth especially designed for superior final polishing. For best results, use with our diamond polishing compound. Pressue Sensitive Adhesive back and 2 1/2" hole.
Type: 2.5" centered hole
Diameter: 8"