Learn More
Electron Microscopy Sciences Kempad-Pan W Polishing Cloth PSA, 2.5" center hole, 8"
SDP
Catalog No. 50997218
Encompass_Preferred
A non-woven textile cloth which is both oil and water resistant. Recommended for intermediate and final polishing with diamond abrasive compounds. Excellent flatness. Available with Pressure Sensitive Adhesive (PSA) with and without hole.
Type: 2.5" centered hole
Diameter: 8"
Catalog No. 50-997-218 Supplier Electron Microscopy Sciences Supplier No. 5031015
A non-woven textile cloth which is both oil and water resistant. Recommended for intermediate and final polishing with diamond abrasive compounds. Excellent flatness. Available with Pressure Sensitive Adhesive (PSA) with and without hole.
Type: 2.5" centered hole
Diameter: 8"