Learn More
Electron Microscopy Sciences Spec-Cloth Spec-Cloth Polishing Cloth, PSA no hole, 8"
SDP
Catalog No. 50997224
Encompass_Preferred
A long napped rayon flock with a plastic barrier. This is a finer version of our Glenco cloth especially designed for superior final polishing. For best results, use with our diamond polishing compound. Pressue Sensitive Adhesive (PSA) back and no hole.
Type: no hole
Diameter: 8"
Catalog No. 50-997-224 Supplier Electron Microscopy Sciences Supplier No. 5032005
A long napped rayon flock with a plastic barrier. This is a finer version of our Glenco cloth especially designed for superior final polishing. For best results, use with our diamond polishing compound. Pressue Sensitive Adhesive (PSA) back and no hole.
Type: no hole
Diameter: 8"