Learn More
Electron Microscopy Sciences Kempad-Pan Kempad-Pan W Polishing Cloth, PSA, No Hole, 8"
SDP
Catalog No. 50997217
Encompass_Preferred
A non-woven textile cloth which is both oil and water resistant. Recommended for intermediate and final polishing with diamond abrasive compounds. Excellent flatness. Available with Pressure Sensitive Adhesive (PSA) with and without hole.
Type: no hole
Diameter: 8"
Catalog No. 50-997-217 Supplier Electron Microscopy Sciences Supplier No. 5031005
A non-woven textile cloth which is both oil and water resistant. Recommended for intermediate and final polishing with diamond abrasive compounds. Excellent flatness. Available with Pressure Sensitive Adhesive (PSA) with and without hole.
Type: no hole
Diameter: 8"