Learn More
EZKEM Polishing Compound CN working Electrode
SDP
Catalog No. NC1813179
Encompass_Preferred
EZkem’s Electrode Polishing Compound is the perfect grade of electrode polishing compound for Silver or Gold Electrodes. Use the electrode polishing compound as required for the normal, ongoing maintenance of the amperometric flowcell when using the Gas Diffusion Cyanide Analysis technique on OI Analytical or various other Autoanalyzer Instrument types.
Catalog No. NC1813179 Supplier EZKEM Supplier No. E060018
EZkem’s Electrode Polishing Compound is the perfect grade of electrode polishing compound for Silver or Gold Electrodes. Use the electrode polishing compound as required for the normal, ongoing maintenance of the amperometric flowcell when using the Gas Diffusion Cyanide Analysis technique on OI Analytical or various other Autoanalyzer Instrument types.