Learn More
Hach Company Polishing kit for Orbisphere sensors - includes 3 μm polishing powder and cloth
Catalog No. NC3269526 Shop All Hach Company Products
Encompass
Polishing kit for sensor surfaces supplied with polishing powder and a polishing cloth. The kit is intended for manual polishing of electrochemical sensor surfaces to restore surface finish, help maintain measurement accuracy, and extend sensor lifetime.
  • Contains 3 μm polishing powder and a polishing cloth.
  • Restores sensor surface finish to improve measurement accuracy.
  • Suitable for field or laboratory manual polishing procedures.
  • Helps extend sensor lifetime when used as part of routine maintenance.
Catalog No. NC3269526 Supplier Hach Company Supplier No. 29781
Polishing kit for sensor surfaces supplied with polishing powder and a polishing cloth. The kit is intended for manual polishing of electrochemical sensor surfaces to restore surface finish, help maintain measurement accuracy, and extend sensor lifetime.
  • Contains 3 μm polishing powder and a polishing cloth.
  • Restores sensor surface finish to improve measurement accuracy.
  • Suitable for field or laboratory manual polishing procedures.
  • Helps extend sensor lifetime when used as part of routine maintenance.