Learn More
Electron Microscopy Sciences Pollack's Trichrome Solution in 50% Alcohol, 3A 100 ML
SDP
Catalog No. 5030102
Encompass_Preferred
Cytoplasmic granule stains and mast cell stains. References: Toren, D.A.: Stain Techn., 38:249-250, 1963. Luna, L., (ed.), Manual of Histologic Staining Methods of the AFIP, 3rd. ed., McGraw-Hill, New York, c. 1968, pp. 116-117.
Catalog No. 50-301-02 Supplier Electron Microscopy Sciences Supplier No. 2647203
Cytoplasmic granule stains and mast cell stains. References: Toren, D.A.: Stain Techn., 38:249-250, 1963. Luna, L., (ed.), Manual of Histologic Staining Methods of the AFIP, 3rd. ed., McGraw-Hill, New York, c. 1968, pp. 116-117.