Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Polo-like kinase 1 protein, human recombinant. | >95.0% | 100 UG
SDP

Catalog No. 5000290317
Encompass_Preferred

Recombinant human polo-like kinase 1 (PLK1) expressed in Sf9 insect cells and supplied as a filtered solution with an N-terminal 10xHis tag. Intended for research applications including biochemical assays, antibody generation, and functional studies of mitosis-related signaling.

  • Expressed in Sf9 insect cells, enabling eukaryotic folding and glycosylation.
  • N-terminal 10xHis tag for affinity purification and detection.
  • Purity >95% by reducing SDS-PAGE.
  • Observed molecular weight ~66-74 kDa due to glycosylation.
  • Supplied as a 0.22 μm filtered solution in glycerol-containing buffer for stability.
  • Store at -80°C; stable at -20°C short term; avoid repeated freeze-thaw.
  • Suitable for kinase assays, antibody production, and cell signaling studies.

Catalog No. 50-002-90317 Supplier Medchemexpress LLC Supplier No. HYP76550100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human polo-like kinase 1 (PLK1) expressed in Sf9 insect cells and supplied as a filtered solution with an N-terminal 10xHis tag. Intended for research applications including biochemical assays, antibody generation, and functional studies of mitosis-related signaling.

  • Expressed in Sf9 insect cells, enabling eukaryotic folding and glycosylation.
  • N-terminal 10xHis tag for affinity purification and detection.
  • Purity >95% by reducing SDS-PAGE.
  • Observed molecular weight ~66-74 kDa due to glycosylation.
  • Supplied as a 0.22 μm filtered solution in glycerol-containing buffer for stability.
  • Store at -80°C; stable at -20°C short term; avoid repeated freeze-thaw.
  • Suitable for kinase assays, antibody production, and cell signaling studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.