Learn More
Medchemexpress LLC Poloxamer 237 (F87) | 9003-11-6 | 7700 (Average) | 500 G
SDP
Supplier:  Medchemexpress LLC HYD1005B500G
Encompass_Preferred
This compound is an alcohol substance that acts as an oil-in-water emulsifier and solubilizer. It is utilized in the research and development of various formulations, including those for ophthalmic applications.
  • Functions as an oil-in-water emulsifier.
  • Acts as a solubilizer.
  • Used in research for contact lens care solutions.
  • Applied in the research of eye drop formulations.
Catalog No. 50-004-29614