Learn More
*CARBOMER INC POLOXAMER 237 NF 100G
Catalog No. NC1346700
Encompass
Poloxamer 237 NF [9003-11-6] Poly(ethylene)mpoly(propylene)n glycol block copolymer m = 64 n = 37 Mw 6,840-8,830 HO(C2H4O)m(C3H6O)n(C2H4O)mH
Catalog No. NC1346700 Supplier *CARBOMER INC Supplier No. 8,01153
Poloxamer 237 NF [9003-11-6] Poly(ethylene)mpoly(propylene)n glycol block copolymer m = 64 n = 37 Mw 6,840-8,830 HO(C2H4O)m(C3H6O)n(C2H4O)mH