Learn More
*CARBOMER INC Poloxamer 338 NF, Poly(ethylene)mpoly(propylene)n glycol block copolymer
Supplier:  *CARBOMER INC 8,01154
Encompass
Poloxamer 338 NF [9003-11-6] Poly(ethylene)mpoly(propylene)n glycol block copolymer m = 141 n = 44 Mw 12,700-17,400 HO(C2H4O)m(C3H6O)n(C2H4O)mH
Catalog No. NC1346705