Learn More
Medchemexpress LLC HY-D1005 500mg Medchemexpress, Poloxamer 407 CAS:9003-11-6 Purity:>98%
SDP
Catalog No. 502082750
Encompass_Preferred
Medchemexpress, HY-D1005 500mg Poloxamer 407 CAS:9003-11-6 Poloxamer 407 is a nonionic surfactant that is 100% active and relatively non-toxic to cells at low concentrations, and frequently used with dye AM esters such as Indo-1 AM, Fura-2 AM, Calcein AM, Fluo-3 AM, Fluo-4 AM, Quest Fluo-8™ AM and Quest Rhod-4™ AM, etc. to improve their water solubility. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2750 Supplier Medchemexpress LLC Supplier No. HYD1005500MG
Medchemexpress, HY-D1005 500mg Poloxamer 407 CAS:9003-11-6 Poloxamer 407 is a nonionic surfactant that is 100% active and relatively non-toxic to cells at low concentrations, and frequently used with dye AM esters such as Indo-1 AM, Fura-2 AM, Calcein AM, Fluo-3 AM, Fluo-4 AM, Quest Fluo-8™ AM and Quest Rhod-4™ AM, etc. to improve their water solubility. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.