Learn More
TARGETMOL CHEMICALS INC POLOXIN-2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T259695MG
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. Poloxin-2 is a potent and selective Plk1 PBD inhibitor with anti-tumour activity that reduces the protein level of Plk1 in HeLa cells. purity: 99%
Catalog No. 50-257-7085