Learn More
Aviva Systems Biology Corp POLR2B Antibody - N-terminal region (ARP32457_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32457_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against POLR2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POLR2B. Alternate names: RPB2, POL2RB, hsRPB2, hRPB140ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54601