Learn More
Aviva Systems Biology Corp POLR2I Antibody - N-terminal region (ARP38758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLR2I. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human POLR2I. Alternate names: RPB9, hRPB14.5ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-56785