Learn More
Aviva Systems Biology Corp POLR3C Antibody - middle region (ARP52190_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52190_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against POLR3C. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human POLR3C. Alternate names: RPC3, RPC62ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-285-657