Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Poltex Benchtop Organizer for HPLC
SDP

Catalog No. 22000245
Change view
Click to view available options
Material:
PETG
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
22-000-245 PETG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-000-245 Supplier Poltex Supplier No. HPLCORG
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Organize HPLC or other lab items (pipettors, transfer pipettes, caps, vials, pipette tips, scissors, pens, etc) on your benchtop in the Poltex Benchtop ORganizer for HPLC. Handle on top for carrying. Made from durable 1/4in thick PETG. Stack items up and out of the way.

Organize HPLC or other lab items (pipettors, transfer pipettes, caps, vials, pipette tips, scissors, pens, etc) on your benchtop. Handle on top for carrying. Made from durable 1/4in thick PETG. Stack items up and out of the way.

Specifications

Autoclavable Not Autoclavable
Color Clear
Material PETG
For Use With (Equipment) HPLC
Length (Metric) 160 mm
Height (Metric) 342 mm
Width (Metric) 323 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.