Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Flex Neck Biohazard Shield with Magnetic Base
SDP

Catalog No. 22000209
Change view
Click to view available options
Length (English):
8.5 in.
12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-000-209 12 in.
22-000-208 8.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-000-209 Supplier Poltex Supplier No. 12FLEXBIOM
  • $263.00
    $227.65 / Each of 1

    Save $35.35

    13% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flexible neck of 12 in. holds firmly in whatever position per user requirements

  • Made from durable scratch and break-resistant polycarbonate
  • Available in two sizes - 8.5 x 12 in. and 12 x 12 in.

Specifications

Dimensions (L x W) 12 x 12 in.
Width (English) 12 in.
Product Type Flexible Neck Biohazard Shield
Includes Magnetic base
Length (English) 12 in.
Material Polycarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.