Learn More
Berkshire™ Polx™ 1200
Catalog No. 0666535ALC Shop All Berkshire Corporation Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
06-665-35ALC Autoclavable
1 options
This item is not returnable. View return policy
Catalog No. 06-665-35ALC Supplier Berkshire™ Supplier No. P1200 0909 8LC
Add to Cart
Add to Cart
This item is not returnable. View return policy
Polx™ 1200 is a knife cut unlaundered cleanroom wiper recommended for ISO Class 5 and above environments.
  • Low contamination compared to non-wovens.
  • High abrasion resistance.
  • Chemically compatible with IPA, Acetone and other solvents.
  • Soft hand and edge for sensitive surfaces.
Specifications
Product Type Absorbent Wipe
Material Polyester
Autoclavable Autoclavable
Disposable Single-use
Ply 1-ply
Sterility Sterile