Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(2,6-dimethyl-1,4-phenylene oxide)

Catalog No. 181781250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-178-1250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-178-1250G Supplier Sigma Aldrich Supplier No. 181781250G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(2,6-dimethyl-1,4-phenylene oxide) (PPO) is an anion exchange membrane (AEM) material that is mainly used as a cross-linker in which the methyl groups are used to bond with ultraviolet (UV) excited ketones.Imidazolium modified PPO with liquid ionic graphene oxide can be used as an anion exchange membrane (AEM) composite with a peak power density of 136 mW cm2 for use in the fabrication of high performance fuel cells. It may also be sulfonated to form a long chained polymeric structure with an efficiency of 81.8% at a current density of 120 mA cm2 for potentiall use in vanadium redox flow batteries (VRFBs) as effective energy storage systems. It may also be used in the formation of AEMs, which can be used in the development of microbial desalination cell system (MDCS).

Specifications

CAS 25134-01-4
Density 1.06 g/mL (at 25°C)
Linear Formula [C6H2(CH3)2 O]n
MDL Number MFCD00084368
Quantity 250 g
Synonym 2,6-Dimethyl-1,4-phenylene ether homopolymer; 2,6-Dimethylphenol homopolymer; 2,6-Dimethylphenol polymer; Poly(2,6-dimethylphenylene oxide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.