Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(2-acrylamido-2-methyl-1-propanesulfonic acid) solution

Catalog No. 191973100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-197-3100G 100 g
19-197-3250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-197-3100G Supplier Sigma Aldrich Supplier No. 191973100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Thickening agent for acidic or basic cleaning agents, friction reducing agent, water-based lubricant, mineral scale remover and suspension aid for pigments and fillers. Rheology control agent in water and some organic solvents.

Specifications

CAS 27119-07-9
Quantity 100 g
MDL Number MFCD00084369
Synonym 2-Acrylamido-2-methyl-1-propanesulfonic acid polymer; 2-Acrylamido-2-methylpropanesulfonic acid polymer; PolyAMPS
Molecular Weight (g/mol) 2000000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.