Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(2-ethylhexyl methacrylate) solution

Catalog No. 18207950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-207-950G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-207-950G Supplier Sigma Aldrich Supplier No. 18207950G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

PEHMA exhibits biomimetic properties. It finds potential application as a flexible hydrogel for ophthalmologic applications.Poly(2-ethylhexyl methacrylate) (PEHMA) may be used to synthesize binary polymeric blends with poly(butyl methacrylate). PEHMA may be utilized in the preparation of PEHMA /carbon black composites for gas sensing. PHEMA-antioxidant conjugate (Quercetin (Q) ) may be prepared for ophthalmologic uses.

Specifications

CAS 25719-51-1
Density 0.908 g/mL (at 25°C)
Linear Formula (C12H22O2)n
Quantity 50 g
MDL Number MFCD00084373
Synonym 2-Ethylhexyl methacrylate homopolymer; Poly(ethylhexyl methacrylate)
Molecular Weight (g/mol) 123000
Biological Activity Central nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.