Learn More
Medchemexpress LLC Poly(2-hydroxyethyl methacrylate) | 25249-16-5 | 1000000 | 250 MG
SDP
Catalog No. 5000439528
Encompass_Preferred
Poly(2-hydroxyethyl methacrylate) (MW 1000000) is recognized as one of the most significant hydrogels within the biomaterials field. It serves as the fundamental component for contact lenses and finds applications in soft tissue implantation, synthetic transplants for cartilage and bone, regeneration of neurotic tissue, and agent transmission, among other uses.
  • Retains moisture content similar to live tissue.
  • Resistant to degradation and absorption by host cells.
  • Can be easily manufactured in various shapes.
  • Readily sterilized.
  • Does not induce cell death in hRPE cultures.
Catalog No. 50-004-39528 Supplier Medchemexpress LLC Supplier No. 250MGHY112531A
Poly(2-hydroxyethyl methacrylate) (MW 1000000) is recognized as one of the most significant hydrogels within the biomaterials field. It serves as the fundamental component for contact lenses and finds applications in soft tissue implantation, synthetic transplants for cartilage and bone, regeneration of neurotic tissue, and agent transmission, among other uses.
  • Retains moisture content similar to live tissue.
  • Resistant to degradation and absorption by host cells.
  • Can be easily manufactured in various shapes.
  • Readily sterilized.
  • Does not induce cell death in hRPE cultures.