Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(2-hydroxyethyl methacrylate)

Catalog No. 19206625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-206-625G 25 g
19-206-610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-206-625G Supplier Sigma Aldrich Supplier No. 19206625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Solid Poly(2-hydroxyethyl methacrylate) (pHEMA) is brittle, while pHEMA hydrogel is a soft material. Glass transition temperature for pHEMA hydrogels is reduced by increasing a content of water in the matrix. Macromolecular properties in pHEMA are characterized by non-covalent interactions of hydrogen-bonds among the polymer chains as well as the hydrated water molecules. The presence of polar groups of hydroxyl and carboxyl on each repeat unit makes this polymer compatible with water. The hydrophobic a-methyl groups of the backbone convey hydrolytic stability to the polymer and enhance mechanical strength of the polymer matrix.pHEMA scaffolds are used in tissue engineering. It can be used in hydrogels for biomedical applications, as sorbents for metal ions.pHEMA has been used in applications such as soft contact lenses, in drug delivery systems, and in kidney dialysis membranes.Nanoparticles of pHEMA may be employed as a carrier for controlled delivery of anticancer and antitumor drugs.

Specifications

CAS 25249-16-5
Density 1.15 g/mL (at 25°C)
Linear Formula (C6H10O3)n
MDL Number MFCD00084374
Quantity 25 g
Synonym Poly(2-HEMA); Poly-HEMA
Molecular Weight (g/mol) 300000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.