Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly[2-methoxy-5-(2-ethylhexyloxy)-1,4-phenylenevinylene]

Catalog No. 541443250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-144-3250MG 250 mg
54-144-31G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54-144-3250MG Supplier Sigma Aldrich Supplier No. 541443250MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

MEH-PPV is a conducting organic semiconductor which has low molecular weight and hydrophobic characteristics. It is a poly(phenylenevinylene) (PPV) derivative and a conjugating polymer with highest occupied molecular orbital (HOMO) below the fermi level of gold.Organic Semiconductor Laser Materials

Specifications

CAS 138184-36-8
Linear Formula (C18H28O2)n
MDL Number MFCD03093943
Quantity 250 mg
Synonym MEH-PPV
Molecular Weight (g/mol) 40,000-70,000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.