Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(3,4-ethylenedioxythiophene)

Catalog No. 67528825ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-528-825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-528-825ML Supplier Sigma Aldrich Supplier No. 67528825ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(3,4-ethylenedioxythiophene)(PEDOT) is a conjugating polymer that has a conductivity of 300 S/cm. It can be copolymerized with poly(styrene-sulfonic acid) to form a water soluble polymeric film with high conductivity and transparency.PEDOT doped with graphene oxide (GO) may be coated on carbon electrodes to fabricate electrochemical sensors for nonenzymatic sensing of glucose. It can also be used as a conductive polymer which may act as a bridge between the electronic polymers and biomolecules for a variety of bioelectronics applications.

Specifications

CAS 126213-51-2
Linear Formula (C6H4O2S)n
Quantity 25 mL
MDL Number MFCD09753886
Synonym PEDOT; Poly(2,3-dihydrothieno-1,4-dioxin)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.