Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(3,4-ethylenedioxythiophene)-block-poly(ethylene glycol) solution

Catalog No. 64980525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-980-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 64-980-525G Supplier Sigma Aldrich Supplier No. 64980525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Please follow this link for Aedotron/Oligotron selection guide table.Nitromethane dispersions can be used to cast thin films on glass, ITO, silicon wafers and many polymeric substrates. Compatibility of the polymeric substrate with nitromethane should be tested in each case. Coat the substrate with a pool of the nitromethane dispersion. Thin transparent films are typically spun at 1000 RPM or higher speeds, but more conducting films can be obtained at lower speeds. A single layer spun at 1000 RPM is typically 75 nm thick.Aedotron is a trademark of TDA Research, Inc.

Specifications

Density 1.127 g/mL (at 25°C)
MDL Number MFCD06798808
Quantity 25 g
Synonym Aedotron(TM) C-NM; PEDOT-block-PEG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.