Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate)

Catalog No. 56059625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-059-625G 25 g
56-059-6100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-059-625G Supplier Sigma Aldrich Supplier No. 56059625G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Preferably applied by spin-coating. Filtration of the dispersion through a 0.45 mum memberane filter is recommended before use. The coatings are dried at a maximum temperature of 200°C for 1 minute, but a temperature between 50°C and 150°C is usually sufficient. The optimal thickness of the dried layer is in the range of 50-250 nm.Virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Conductive polymer blend.

Specifications

Quantity 25 g
MDL Number MFCD07371079
Synonym PEDOT:PSS; Poly(2,3-dihydrothieno-1,4-dioxin)-poly(styrenesulfonate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.