Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Poly 30 Gal OH drum with Plastic Ring; Red
SDP

Catalog No. 19339775
Change view
Click to view available options
Capacity (English):
30 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-339-775 30 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-339-775 Supplier Youngstown Barrel & Drum Supplier No. 1556MR

missing translation for 'validMainframeMsg'

Constructed of red blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, weather and chemical resistant, and has a white lid with a plastic lever-lock band closure. Ideal for safe storage, handling and transportation of hazardous materials and spill containment.

Specifications

Certifications/Compliance DOT 49 CFR 173.12
Color Red
Capacity (English) 30 gal.
Capacity (Metric) 113.6 L
Material Polyethylene
Type Open Head Drum with Poly Ring
For Use With (Application) safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration. disposal and incineration.
Includes Lid and ring
Model Open Head Drum
Packaging Quantity 1000/Pk.
Resistant Chemical, Weather
Shape Circular
Dimensions (L x W x H) 21 1/8 x 21 1/8 x 28 1/2 in.
Width (English) 21-1/8 in.
Width (Metric) 53.34 cm
Height (English) 28 1/2 in.
Height (Metric) 72.39 cm
Diameter (English) 21-1/8 in. Top, 16-5/8 in. Bottom
Diameter (Metric) Top: 54 cm, Bottom: 42.2 cm
Weight (English) 18 lb.
Weight (Metric) 8.16 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.