Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly[4,5-difluoro-2,2-bis(trifluoromethyl)-1,3-dioxole-co-tetrafluoroethylene]

Catalog No. 4696291G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-962-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46-962-91G Supplier Sigma Aldrich Supplier No. 4696291G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(4,5-difluoro-2,2-bis(trifluoromethyl)-1,3-dioxole-co-tetrafluoroethylene)(AF2400) is a fluorinated polymer that formed by copolymerization of 4,5-difluoro-2,2-(trifluoromethyl)-1,3-dioxole(PDD) and tetrafluoroethylene(TFE). It has a relatively high glass transition temperature(Tg) and optical transparency which allows it be a photo-stable polymer that can be used for a wide range of coating application in electronic and electrochemical industry.Optoelectronic devices for optical clarity, low-refractive index coating for optical devices, protective coating for chemical resistance, release coating, and sight window for harsh chemical environments.Teflon is a registered trademark of E. I. du Pont de Nemours and Company

Specifications

CAS 37626-13-4
Density 1.67 g/mL (at 25°C)
Refractive Index n20/D 1.29
MDL Number MFCD00803821
Quantity 1 g
Synonym PTFE AF 2400 PTFE AF; PTFE™ AF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.