Learn More
Sino Biological Peptide Substrates: Poly (4:1 Glu, Tyr) Peptide
Catalog No. 502568152
Encompass_Preferred
The Poly (4:1 Glu, Tyr) substrate peptide is used as universal substrate for protein tyrosine kinases.
Catalog No. 50-256-8152 Supplier Sino Biological Supplier No. P6158
The Poly (4:1 Glu, Tyr) substrate peptide is used as universal substrate for protein tyrosine kinases.