Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly[9,9-bis-(2-ethylhexyl)-9H-fluorene-2,7-diyl]

Catalog No. 5710321G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-103-21G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-103-21G Supplier Sigma Aldrich Supplier No. 5710321G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly[9,9-bis-(2-ethylhexyl)-9H-fluorene-2,7-diyl] (PFO) is a polyfluorene based conductive liquid crystalline material that can be used as a light emitting material. It forms a semiconducting layer to enhance the electroluminescence (EL) and performance of the photonic and optoelectronic devices.PFO forms aconjugating polymeric system that can produce blue EL devices which include organic light emitting diodes (OLEDs), optical fibers and molecular electronic devices.

Specifications

CAS 188201-16-3
MDL Number MFCD03703236
Quantity 1 g
Synonym Poly[9,9-di-(2 -ethylhexyl)fluorenyl-2,7-diyl]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.