Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(9-vinylcarbazole)

Catalog No. 1826055G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-260-55G 5 g
18-260-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-260-55G Supplier Sigma Aldrich Supplier No. 1826055G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(9-vinylcarbazole) (PVK) is a conductive polymer which is mainly used as a hole transporting medium at high efficiencies with low driving voltage. It can also be used as an anode for hole injection and can act as an effective charge transferring gate by co-doping it with organic dyes.PVK:graphene oxide composite can be used as a hybrid film which can be coated on silica substrate for the fabrication of organic field effect transistors (OFETs). It can also form a nano-composite film with carbon nanospheres (CNS) which can be potentially used in supercapacitors and sensor based applications.

Specifications

CAS 25067-59-8
Density 1.2 g/mL (at 25°C)
Refractive Index n20/D 1.683
MDL Number MFCD00134336
Quantity 5 g
Synonym PVK
Molecular Weight (g/mol) 1100000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.