Learn More
Poly(acrylic acid), 25 wt% soln. in water
Description
Polyacrylic acid is used in disposable diapers and in ion exchange resins. It is also used to study solute diffusion in polyvinyl alcohol/polyacrylic acid copolymer hydrogel. It is also employed as a thickening, suspending, emulsifying and dispersing agent in pharmaceuticals, cosmetics, adhesives and paints. Further, it is used for the preparation of poly(N-isopropylacrylamide)-block-polyacrylic acid copolymer which responds to both temperature and pH stimuli. In addition to this, it is used in preparing block copolymer of oligo (methyl methacrylate)/PAA for micellar delivery of hydrophobic drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
            Specifications
Specifications
| Physical Form | Liquid | 
| Density | 1.09 g/mL | 
| Solubility Information | Soluble in water. | 
| Quantity | 100 g | 
| Concentration or Composition (by Analyte or Components) | 25 wt% soln. in water | 
| Chemical Name or Material | Poly (acrylic acid) | 
Frequently Asked Questions (FAQs)
The average molecular weight of the Poly(acrylic acid), 25 wt% soln. in water is 240,000 g/mole.
Safety and Handling
| RTECSNumber | AT4680000 | 
| TSCA | Yes | 
| Recommended Storage | Ambient temperatures | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.